Learn More
ATCC Salmonella enterica; subsp. enterica Poona
Supplier:  ATCC BAA3139
Encompass
Salmonella enterica subsp. enterica serovar Poona is a whole-genome sequenced strain that was isolated in 2010 from fresh cucumber in the Dominican Republic. It has applications in enteric, infectious, and zoonotic disease research.
Catalog No. 50-238-2624