Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Typhimurium 6 x 0.3ml
Supplier:  ATCC 13311MINIPACK
Encompass
ATCC 13311-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 13311™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-4686