Learn More
ATCC Salmonella enterica; subsp. enterica; serovar typhimurium
Supplier:  ATCC 29629
Encompass
Salmonella enterica subsp. enterica strain TA 1535 is a whole-genome sequenced bacterium with applications in enteric disease research, infectious disease research, and zoonotic disease research. This strain is serotype I 4,5,12:i:1,2 and is the HisG46, rfa-, uvrB derivative of strain LT-2.
Catalog No. 50-238-4786