Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Choleraesuis
Supplier:  ATCC 10708
Encompass
Salmonella enterica subsp. enterica serovar Choleraesuis strain ETS 34 is a whole-genome sequenced bacterial pathogen with applications in enteric and zoonotic disease research. This strain is serotype I 6,7:c:1,5 and is recommended by ATCC for use in the tests described in Military Spec. MIL G-13734B where only the taxon is specified.
Catalog No. 50-238-5117