Learn More
ATCC Salmonella enterica subsp. enterica serovar Heidelberg (ATCC 8326) - Freeze-dried research strain
Supplier:  ATCC ATCC8326
Encompass
Salmonella enterica subsp. enterica serovar Heidelberg (ATCC 8326™) is a freeze-dried bacterial reference strain intended for laboratory research in enteric and infectious diseases. The strain is supplied as a deposited culture with supporting genomic data and should be handled using BSL-2 safety practices.
  • Freeze-dried format for long-term storage.
  • Aerobic growth; incubate at 37 °C.
  • Recommended media: nutrient agar or nutrient broth.
  • Store freeze-dried vials at 2-8 °C.
  • Biosafety level 2 organism; handle in a BSL-2 facility.
  • Provided as a characterized reference strain with genomic data available.
Catalog No. NC3918087