Learn More
ATCC Salmonella enterica; subspecies enterica; serovar Typhimurium; Strain: SAP17-8290
Supplier:  ATCC BAA3157
Encompass
Salmonella enterica subsp. enterica serovar Typhimurium strain SAP17-8290 was isolated in the United States from human feces. This whole-genome sequenced bacterium was part of the 2017 PulseNet-GenomeTrakr joint proficiency test exercise. This strain has applications in infectious disease research and zoonotic disease research.
Catalog No. 50-238-2567