Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Typhimurium; phage P22
Supplier:  ATCC 19585B1
Encompass
Bacteriophage P22 is propagated in Salmonella enterica subsp. enterica serovar Typhimurium strain LT2. This organism has applications as a transducing phage for Salmonella Groups B and D.
Catalog No. 50-238-3330
 Restricted Item