Learn More
Medchemexpress LLC Salsolidine | 5784-74-7 | 97.4% | 207.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY2238550MG
Encompass_Preferred
Salsolidine is a tetrahydroisoquinoline alkaloid that acts as a stereoselective competitive MAO A inhibitor. It is intended for research use only.
  • Tetrahydroisoquinoline alkaloid
  • Stereoselective competitive MAO A inhibitor
  • R-salsolidine is more active against MAO A than S-salsolidine (Ki=6 μM and 186 μM, respectively)
  • Weakly inhibits binding of δ-receptor (Ki >100 μM)
  • Potential for inhibiting Acetylcholinesterase and butyrylcholinesterase
  • Appears as a solid
  • Color is light yellow to brown
  • Initial source is animals (bovine liver)
Catalog No. 50-004-02577