Learn More
Thermco Products Inc Salt Brine Hydrometer | 0-100%
Supplier:  Thermco Products Inc GW2511
Encompass_Preferred
Salt Brine Hydrometers (Salometers) measure the saturation of salt in an aqueous solution.
Catalog No. NC1709534