Learn More
New England Biolabs, Inc. Salt-T4 DNA Ligase – 20000 units
SDP
Supplier:  New England Biolabs, Inc. M0467S
Encompass
A salt tolerant variant of T4 DNA Ligase, Salt-T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA and is designed to function at higher salt concentrations than wild type T4 DNA Ligase. This enzyme will join cohesive end termini at salt concentrations as high as 300 mM without any loss in activity. This enzyme is insensitive to salt carried over from other reaction components (vector or insert DNA) and allows ligation reactions to proceed in alternative reaction buffers with higher levels of salt (e.g., NEBuffer r3.1).
Catalog No. 50-202-4037