Learn More
Hampton Research HR2-909-04/SaltRx 2 #04 (E04), 185 ml - 2.4 M Ammonium phosphate dibasic, 0.1 M Tris pH 8.5
SDP
Supplier:  Hampton Research HR290904
Encompass
HR2-909-04/SaltRx 2 #04 (E04), 185 ml - 2.4 M Ammonium phosphate dibasic, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-136-5