Learn More
Hampton Research SALTRX 2 NO16 185ML
SDP
Supplier:  Hampton Research HR290916
Encompass
HR2-909-16/SaltRx 2 #16 (F04), 185 ml - 2.5 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-137-7