Learn More
Hampton Research SALTRX 2 NO20 185ML
SDP
Supplier:  Hampton Research HR290920
Encompass
HR2-909-20/SaltRx 2 #20 (F08), 185 ml - 0.8 M Lithium sulfate monohydrate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-138-1