Learn More
Hampton Research SALTRX 2 NO22 185ML
SDP
Supplier:  Hampton Research HR290922
Encompass
HR2-909-22/SaltRx 2 #22 (F10), 185 ml - 1.5 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-138-3