Learn More
Hampton Research SALTRX 2 NO24 185ML
SDP
Supplier:  Hampton Research HR290924
Encompass
HR2-909-24/SaltRx 2 #24 (F12), 185 ml - 1.5 M Lithium sulfate monohydrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-138-5