Learn More
Hampton Research SALTRX 2 NO32 185ML
SDP
Supplier:  Hampton Research HR290932
Encompass
HR2-909-32/SaltRx 2 #32 (G08), 185 ml - 0.7 M Ammonium tartrate dibasic, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-139-3