Learn More
Hampton Research SALTRX 2 NO37 185ML
SDP
Supplier:  Hampton Research HR290937
Encompass
HR2-909-37/SaltRx 2 #37 (H01), 185 ml - 0.6 M Potassium sodium tartrate tetrahydrate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-139-8