Learn More
Hampton Research SALTRX 2 NO39 185ML
SDP
Supplier:  Hampton Research HR290939
Encompass
HR2-909-39/SaltRx 2 #39 (H03), 185 ml - 0.6 M Potassium sodium tartrate tetrahydrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-140-0