Learn More
Hampton Research SALTRX 2 NO43 185ML
SDP
Supplier:  Hampton Research HR290943
Encompass
HR2-909-43/SaltRx 2 #43 (H07), 185 ml - 0.5 M Potassium thiocyanate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-140-4