Learn More
Hampton Research SALTRX 2 NO46 185ML
SDP
Supplier:  Hampton Research HR290946
Encompass
HR2-909-46/SaltRx 2 #46 (H10), 185 ml - 4.0 M Ammonium acetate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-140-7