Learn More
Hampton Research SALTRX 2 NO47 185ML
SDP
Supplier:  Hampton Research HR290947
Encompass
HR2-909-47/SaltRx 2 #47 (H11), 185 ml - 35 % v/v Tacsimate pH 7.0, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-140-8