Learn More
Selleck Chemical LLC Salubrinal 5mg 405060-95-9
SDP
Supplier:  Selleck Chemical LLC S29235MG
Encompass_Preferred
Salubrinal is a selective inhibitor of eIF2? dephosphorylation and inhibits ER stress-mediated apoptosis with EC50 of ~15 ?M in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3220