Learn More
Cayman Chemical Salva plba . B. upafoln 10mg
SDP
Supplier:  Cayman Chemical 4013810 MG
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits iron-dependent lipid peroxidation in a cell-free assay (IC50 = 7.9 µM); inhibits PrP106-126 aggregation in a cell-free assay and reduces PrP106-126-induced cell death in SH-SY5Y cells from 50-150 µM; inhibits the production of hemolysin and leukocidin by MSSA and MRSA and reduces S. aureus-induced hemolysis of erythrocytes in isolated rabbit whole blood at 100 µM; reduces the number of lung CFUs and increases survival in a mouse model of MRSA infection at 100 mg/kg; reduces angiogenesis and tumor volume in a Hep3B mouse xenograft model at 60 mg/kg
Catalog No. 50-294-2141