Learn More
Medchemexpress LLC Salvianolic acid Y | 1638738-76-7 | 1 MG

Supplier: Medchemexpress LLC HYN78801MG
Salvianolic acid Y is a phenolic acid that shares the same planar structure as Salvianolic acid B. It is noted for its ability to rescue cell injury caused by H2O2. This product is for research use only.
- Purity: 99.37%
- Appearance: Solid
- Color: White to light yellow
- Shows protection activity against hydrogen peroxide (H2O2)-induced cell lesion
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.