Learn More
Cell Biolabs Inc S-Adenosylmethionine (SAM) and S-Adenosylhomocysteine (SAH) ELISA Combo Kit, 96 assays
Supplier:  Cell Biolabs Inc MET5151C
Encompass_Preferred
S-Adenosylmethionine (SAM) and S-Adenosylhomocysteine (SAH) are regulators of DNA methylation. SAM can donate a methyl group to DNA; this process converts SAM to SAH, which is an inhibitor of methylation. Cell Biolabs' S-Adenosylmethionine (SAM) and S-Adenosylhomocysteine (SAH) ELISA Combo Kit allows you to quantify both SAM and SAH in a convenient 96-well microplate format. The kit can detect free SAM levels as low as 0.625 µM and free SAH levels as low as 0.2 µM. SAM and SAH are measured in separate wells of the 96-well plate. Each kit provides sufficient reagents for a total of 96 wells to test either SAM or SAH, including standards and unknown samples.
Catalog No. NC1976624