Learn More
Medchemexpress LLC Samidorphan | 852626-89-2 | 99.0% | 370.44 g·mol⁻¹ | C21H26N2O4 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1236895MG
Encompass_Preferred
Samidorphan is an opioid receptor modulator used in research; it acts as an orally active opioid system modulator with affinity for mu, kappa, and delta opioid receptors. The compound is supplied as a solid (white to yellow), with molecular formula C21H26N2O4 and molecular weight 370.44 g·mol⁻¹, and is intended for preclinical and in vitro studies.
Catalog No. 50-002-37529