Learn More
Perkin Elmer US LLC Sample Capillary for ST Nebulizers - 1.0 mm I.D., 1 mL/min, Gray
Supplier:  Perkin Elmer US LLC N8145138
Encompass_Preferred
This Sample Capillary, marked gray, has a self-aspiration rate of 1 mL/min at 1 L/min of Argon flow.
Catalog No. 50-210-4924