Learn More
World Precision Instrument Sample Injector Assembly
SDP
Supplier:  World Precision Instrument 58006
Encompass
Fluid pressure fluctuation, cross-contamination and the introduction of small air bubbles in a liquid waveguide capillary cell (LWCC) can cause baseline variations and measurement inaccuracies. Use the optional Sample Injector Kit (WPI #58006) or the LWCC Injection System (WPI #89372) to minimize the development of small air bubbles.
Catalog No. 50-252-1245