Learn More
Hach Company External Pump (sample leveling) Complete, for AT1000 Series Titrator
Supplier:  Hach Company LZE142
Encompass
The external peristaltic pump comes complete with tubing and connecting cable. This AT1000 optional accessory move one step further the analysis automation performing an automatic sample amount adjustment by removing sample in excess avoiding operator to do sample pipetting. Sample leveling is preprogrammed in applications delivered in our packs for liquid samples.
Catalog No. 50-199-7876