Learn More
Research Products International Corp Micro-Tube Sample Pestles with 1.5 ml Micro-Tubes, RNase and DNase Free, 50 Sets per Case
SDP
Supplier:  Research Products International Corp 199226
Encompass
Ideal for manually homogenizing cells and resuspending proteins or DNA pellets in buffers. Pestles feature conical tip with matched tube for maximum sample contact. Pestles and micro-tubes molded from polypropylene and can be autoclaved. Supplied in assorted colors: Red, Blue, Green, Purple, Orange.
Catalog No. 50-550-226