Learn More
Chemglass Life Sciences Sample Rack for CG-1991-V-133 (For COD analysis only)
SDP
Supplier:  Chemglass Life Sciences CG1991V205
Encompass_Preferred
Sample Rack for CG-1991-V-133 For COD analysis only. For use with CG-1991-V ECODryBlock Systems / Dryblock Heaters.
Catalog No. 50-004-93073