Learn More
Chemglass Life Sciences Syringe, Sample Retrieval, Minum-Ware
SDP
Supplier:  Chemglass Life Sciences MW13201
Encompass_Preferred
Syringe, Sample Retrieval, Minum-Ware. An all borosilicate glass sample syringe with 1ml capacity The stainless steel luer lock assures use of syringe with needle “blow-off”. Needle sold separately, see CG-3075.
Catalog No. 50-872-586