Learn More
Cygnus Technologies Cygnus Technologies F402 Sample Treatment Plate, For ELISA Assays, Includes Adhesive Seal, 1 Plate
Supplier:  Cygnus Technologies F402
Encompass
The Cygnus Technologies Sample Treatment Plate is essential for efficient sample preparation in Protein A ELISA assays. This product facilitates accurate sample dilutions, ensuring reliable detection of impurities. Each unit includes one plate and a foil adhesive seal, and its compatibility with multiple Protein A ELISA Kits differentiates it from other offerings, streamlining the sample treatment process.
Catalog No. NC1629851