Learn More
METTLER TOLEDO 3P INDUSTRIAL Sample Vials, 10ml, Pack of 192
Supplier:  METTLER TOLEDO 3P INDUSTRIAL 30407445
Encompass_Preferred
10mL Sample Vials are a pack of 192 clear glass vials. They have an overall height of 45mm, outer diameter of 14mm, and opening diameter of 7mm. The vials are made of hydrolytic class 1 borosilicate glass and can withstand temperatures from -20°C to 121°C. These vials are suitable for sample storage and analysis in a variety of laboratory applications. Does not include caps.
Catalog No. NC1506719