Learn More
Hach Company Sampler, Dipper, Economy, 6ft
Supplier:  Hach Company 2929302
Encompass
For convenient sampling from tanks, vats, streams, ponds, lakes, etc., these economy dippers are light, easy to use and portable. Two pouring spouts on the 500 mL (16 oz) dipper make it useful for left or right-handed people to pour from either side. Ends are sealed for easy cleaning and include a hook for hanging. The polypropylene cups have threaded fittings that screw onto the handle and are autoclavable at 121ºC (250ºF).
Catalog No. 50-199-6826