Learn More
BURKLE INC Sampling lance Milky, V4A, 18.90 in. (480 mm), 5.07 oz. (150 ml), Ø 1.10 in. (28 mm)
Supplier:  BURKLE INC 53210300
Encompass_Preferred
Universal powder and granules sampler made of stainless steel AISI 316L (1.4404).
Developed initially for milk powder samples, conforms to standards of DIN and IDF (International Dairy Federation).
The collector is inserted into bags or casks and the sample is removed using a semi-circular profile lance.
Catalog No. NC2187682