Learn More
Medchemexpress LLC Samuraciclib hydroch 10mM 1mL | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY103712B10MM 1ML
Encompass_Preferred
Samuraciclib (CT7001) hydrochloride hydrate is a potent selective ATP-competitive and orally active CDK7 inhibitor with an IC50 of 41 nM Samuraciclib hydrochloride hydrate displays 45- 15- 230- and 30-fold selectivity over CDK1 CDK2 (IC50 of 578 nM) CDK5 and CDK9 respectively Samuraciclib hydrochloride hydrate inhibits the growth of breast cancer cell lines with GI50 values between 0 2-0 3 M Samuraciclib hydrochloride hydrate has anti-tumor effects[1 [2
Catalog No. 50-002-44199