Learn More
Electron Microscopy Sciences A microscope slide that has a 3/4" sanblasted , frosted end. Size: 25 x 75 mm x 1 mm (T)
SDP
Supplier:  Electron Microscopy Sciences 7187719
Encompass_Preferred
A microscope slide that has a 3/4" sanblasted , frosted end. Size: 25 x 75 mm x 1 mm (T)
Catalog No. NC3849749