Learn More
Medchemexpress LLC Sangivamycin | 18417-89-5 | 97.7% | 309.28 | C12H15N5O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11838450MG
Encompass_Preferred
Sangivamycin is a nucleoside analog provided as an analytical/reference standard for research use with reported protein kinase C (PKC) inhibitory activity. The compound is offered in solid and solution formats and is supplied in a 50 MG pack; molecular formula C12H15N5O5, molecular weight 309.28, listed purity ≈97.68%.
  • Analytical/reference standard for research and assay use.
  • Nucleoside analog with reported PKC inhibitory activity.
  • Available in solid and solution formats, including 50 MG pack.
  • Listed purity approximately 97.68%.
  • Molecular weight 309.28, formula C12H15N5O5.
Catalog No. 50-002-27180