Learn More
Selleck Chemical LLC Sanguinarine chloride S5452-1mg
SDP
Supplier:  Selleck Chemical LLC S54521MG
Encompass_Preferred
Sanguinarine a plant alkaloid is a potent and specific protein phosphatase (PP) 2C inhibitor Sanguinarine chloride can stimulate apoptosis via activating the production of reactive oxygen species (ROS) Sanguinarine-induced apoptosis is associated with the activation of JNK and NF- B
Catalog No. 50-311-0688