Learn More
Medchemexpress LLC Sanguisorbigenin | 6812-98-2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN815110MG
Encompass_Preferred
Sanguisorbigenin is a natural antibacterial agent that inhibits methicillin-resistant S. aureus (MRSA).
  • Minimal inhibitory concentration values against six strains of S. aureus are in the range of 12.5-50 μg/mL.
  • Significantly inhibits the expression of blaR1, mecA, and blaZ in MRSA in a dose-dependent manner.
  • Effectively reverses antibiotic resistance by inhibiting mecA expression and decreasing PBP2a expression.
Catalog No. 50-004-17184