Learn More
Grainger Sanitary Conductivity Sensor SS
Supplier:  Grainger 16Z059
Encompass_Preferred
The SensoRex Toroidal Conductivity Sensor is an industrial sensor designed for continuous monitoring of conductivity in liquids. It features a toroidal measuring cell made of PEEK with platinum electrodes, providing corrosion resistance. With a measuring range of 0-1000 mS/cm, it outputs a 4-20mA signal proportional to conductivity. The sensor has a max pressure rating of 17 bar at 20°C and is ideal for CIP/SIP applications. With its hygienic design, it meets 3-A Sanitary Standards.
Catalog No. 50-276-8164