Learn More
Essendant SANITIZERHAND55GALCLR
Supplier:  Essendant DVO101104120
Encompass_Preferred
Alcohol based instant hand sanitizer enriched with skin conditioning emollients to keep skin soft and protect against irritation for even the most frequent users Fragrance-free and dye-free Contains 76% (v/v) 72% (w/w) ethyl alcohol Meets CDC WHO APIC and PHAC hand hygiene recommendations for healthcare personnel hand rubs
Catalog No. 50-000-23370