Learn More
Enzo Life Sciences SANT-2 (1mg). CAS: 329196-48-7
SDP
Supplier:  Enzo Life Sciences ALX270436M001
Encompass_Preferred
Cell permeable antagonist of the Smoothened (Smo) protein function. Inhibitor of SAG (ALX-270-426) activity. Chemical tool to study the Hedgehog (Hh) signalling pathway and to explore therapeutic uses for inhibitors of Smo. Alternative name: N-[3-(1H-Benzimidazol-2-yl)-4-chlorophenyl]-3,4,5-triethoxybenzamide. Purity: ≥95%. Appearance: White to off-white solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8557