Learn More
Cayman Chemical SapacItabIn 5mg
SDP
Supplier:  Cayman Chemical 273215 MG
Encompass_Preferred
A prodrug form of CNDAC; reduces tumor volume in an MV4-11 mouse xenograft model at 5 and 15 mg/kg alone or in combination with SAHA
Catalog No. 50-293-9003