Learn More
TARGETMOL CHEMICALS INC SAPANISERTIB 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T183825MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sapanisertib (INK 128) is an orally bioavailable inhibitor of raptor-mTOR (TOR complex 1 or TORC1) and rictor-mTOR (TOR complex 2 or TORC2) with potential antineoplastic activity. purity: 99%
Catalog No. 50-258-1081