Learn More
Medchemexpress LLC HY-13328 50mg , Sapanisertib CAS:1224844-38-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1332850MG
Encompass_Preferred
HY-13328 50mg Sapanisertib CAS: 1224844-38-5 Sapanisertib (INK-128; MLN0128; TAK-228) is an orally available, ATP-dependent mTOR1/2 inhibitor with an IC50 of 1 nM for mTOR kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9698