Learn More
Selleck Chemical LLC Sapitinib (AZD8931) 5mg 848942-61-0
SDP
Supplier:  Selleck Chemical LLC S21925MG
Encompass_Preferred
Sapitinib (AZD8931) is a reversible, ATP competitive inhibitor of EGFR, ErbB2 and ErbB3 with IC50 of 4 nM, 3 nM and 4 nM in cell-free assays, more potent than Gefitinib or Lapatinib against NSCLC cell, 100-fold more selective for the ErbB family than MNK1 and Flt. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1359786