Learn More
Selleck Chemical LLC Saponaria Officinalis Extract E3193-5MG
SDP
Supplier:  Selleck Chemical LLC E31935MG
Encompass_Preferred
Saponaria Officinalis Extract is extracted from Saponaria Officinalis which reduces downstream inflammatory responses by disrupting the TLR4/MD2 complex and blocking MyD88-dependent inflammatory signaling
Catalog No. 50-000-40432