Learn More
Crescent Chemical Co Inc Saponin, 8047-15-2, MFCD00081981, 50g
SDP
Supplier:  Crescent Chemical Co Inc SE34655.01
Encompass_Preferred
Saponin, 8047-15-2, MFCD00081981, 50g, pure, DAB, Isolated from Quillaja bark. For permeabilization of cells. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-112